4-Fluoro-m-toluic acid 97%

Supplier: Ambeed
Warning

A216914-100G A216914-10G A216914-1G A216914-25G A216914-5G
76975-710EA 81.69 USD
76975-710 76975-712 76975-714 76975-716 76975-718
4-Fluoro-m-toluic acid 97%
4-Fluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD01631424
CAS Number: 403-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR