2-Amino-N-[2-(dimethylamino)ethyl]-2-methylpropanamide dihydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-N-(2-(dimethylamino)ethyl)-2-methylpropanamide dihydrochloride

A407490-100MG A407490-1G A407490-250MG A407490-5G
76979-124EA 18.04 USD
76979-124 76979-126 76979-128 76979-130
2-Amino-N-[2-(dimethylamino)ethyl]-2-methylpropanamide dihydrochloride 98+%
2-Amino-N-[2-(dimethylamino)ethyl]-2-methylpropanamide dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₂₁Cl₂N₃O
MW: 246.18 g/mol
MDL Number: MFCD13562564
CAS Number: 1219957-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR