2-Bromo-3-fluoropyridine 98%

Supplier: Ambeed
Warning

A754397-100G A754397-10G A754397-1G A754397-25G A754397-5G
76981-388EA 74 USD
76981-388 76981-390 76981-392 76981-394 76981-396
2-Bromo-3-fluoropyridine 98%
2-Bromo-3-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrFN
MDL Number: MFCD04114128
CAS Number: 40273-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR