2-Iodo-3-pyridinol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-2-iodopyridine

A220376-100G A220376-10G A220376-1G A220376-25G A220376-5G
76975-812EA 252.5 USD
76975-812 76975-814 76975-816 76975-818 76975-820
2-Iodo-3-pyridinol 97%
2-Iodo-3-pyridinol

Precaución: for Research Use Only

Formula: C₅H₄INO
MW: 221.00 g/mol
MDL Number: MFCD00023421
CAS Number: 40263-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR