L-Ornithine methyl ester dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Orn-OMe.2HCl, H-Orn-OMe·2 HCl, (S)-methyl 2,5-Diaminopentanoate dihydrochloride, H-Orn-OMe·2HCl
A107954-100G
A107954-10G
A107954-25G
A107954-5G
76970-546EA
51.99
USD
76970-546
76970-548
76970-550
76970-552
L-Ornithine methyl ester dihydrochloride 97%
L-Ornithine methyl ester dihydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₆Cl₂N₂O₂ MW: 219.106 g/mol |
MDL Number:
MFCD00190977 CAS Number: 40216-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...