Boc-N-Me-Phe-OH·DCHA 98+%

Supplier: Ambeed
Warning

A332794-100G A332794-1G A332794-25G A332794-5G
76978-284EA 197 USD
76978-284 76978-286 76978-288 76978-290
Boc-N-Me-Phe-OH·DCHA 98+%
Boc-N-Me-Phe-OH·DCHA

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄·C₁₂H₂₃N
MDL Number: MFCD00070397
CAS Number: 40163-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR