(Methylthio)acetaldehyde dimethyl acetal 98%

Supplier: Ambeed
Danger

Synonyms: (Methylthio)acetaldehydedimethylacetal

A439494-25G A439494-5G
76979-354EA 75.33 USD
76979-354 76979-356
(Methylthio)acetaldehyde dimethyl acetal 98%
(Methylthio)acetaldehyde dimethyl acetal

Precaución: for Research Use Only

Formula: C₅H₁₂O₂S
MW: 136.21 g/mol
MDL Number: MFCD00009848
CAS Number: 40015-15-4
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR