5-Amino-2-methylpyrimidine 97%

Supplier: Ambeed
Warning

A148505-1G A148505-250MG A148505-5G
76972-980EA 53.05 USD
76972-980 76972-982 76972-984
5-Amino-2-methylpyrimidine 97%
5-Amino-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.1300 g/mol
MDL Number: MFCD09608109
CAS Number: 39889-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR