4-Bromo-2-nitro-5-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-2-bromo-4-nitrobenzotrifluoride

A259904-1G A259904-250MG A259904-5G
76976-902EA 22 USD
76976-902 76976-904 76976-906
4-Bromo-2-nitro-5-(trifluoromethyl)aniline 97%
4-Bromo-2-nitro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄BrF₃N₂O₂
MW: 285.0181 g/mol
MDL Number: MFCD09263997
CAS Number: 683241-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR