2-Amino-4-hydroxy-6-methylpyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-methylpyrimidin-4(1H)-one, 2-Amino-6-methyl-4-pyrimidinol

A113045-1000G A113045-100G A113045-10G A113045-25G A113045-500G
76970-970EA 270.53 USD
76970-970 76970-972 76970-974 76970-976 76970-978
2-Amino-4-hydroxy-6-methylpyrimidine 97%
2-Amino-4-hydroxy-6-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₇N₃O
MW: 125.13 g/mol
MDL Number: MFCD00006095
CAS Number: 3977-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR