3-Ethylcyclohexanone 95% (by GC)

Supplier: Ambeed

A801556-1G A801556-250MG A801556-5G
76981-666EA 149 USD
76981-666 76981-668 76981-670
3-Ethylcyclohexanone 95% (by GC)
3-Ethylcyclohexanone

Precaución: for Research Use Only

Formula: C₈H₁₄O
MDL Number: MFCD13562924
CAS Number: 22461-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR