2-Amino-4-bromo-5-(trifluoromethyl)phenylamine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-5-(trifluoromethyl)benzene-1,2-diamine

A545553-100MG A545553-1G A545553-250MG
76980-208EA 90.18 USD
76980-208 76980-210 76980-212
2-Amino-4-bromo-5-(trifluoromethyl)phenylamine 98%
2-Amino-4-bromo-5-(trifluoromethyl)phenylamine

Precaución: for Research Use Only

Formula: C₇H₆BrF₃N₂
MW: 255.04 g/mol
MDL Number: MFCD09263998
CAS Number: 157590-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR