5-Bromo-3-fluoro-4-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A224703-100MG A224703-10G A224703-1G A224703-250MG A224703-25G A224703-5G
76975-944EA 12 USD
76975-944 76975-946 76975-948 76975-950 76975-952 77881-571
5-Bromo-3-fluoro-4-methylbenzaldehyde 98%
5-Bromo-3-fluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD18205969
CAS Number: 1370411-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR