2-Chloro-5-fluorophenylacetonitrile 97%

Supplier: Ambeed
Warning

A182504-10G A182504-1G A182504-250MG A182504-25G A182504-5G
76974-490EA 60 USD
76974-490 76974-492 76974-494 76974-496 76974-498
2-Chloro-5-fluorophenylacetonitrile 97%
2-Chloro-5-fluorophenylacetonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MDL Number: MFCD06246270
CAS Number: 395675-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR