3-Methoxy-5-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

A244951-5G A244951-1G A244951-250MG
76976-556EA 42 USD
76976-556 77672-190 77881-650
3-Methoxy-5-(trifluoromethyl)benzoic acid 98%
3-Methoxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
MDL Number: MFCD09025409
CAS Number: 53985-48-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR