7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinazoline 95%

Supplier: Ambeed
Warning

A120657-100MG A120657-1G A120657-250MG
76971-492EA 118 USD
76971-492 76971-494 76971-496
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinazoline 95%
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinazoline

Precaución: for Research Use Only

Formula: C₁₄H₁₇BN₂O₂
MW: 256.10 g/mol
MDL Number: MFCD18205965
CAS Number: 1375108-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR