2-Bromo-6-fluoropyrazine 97%

Supplier: Ambeed
Danger

A167359-250MG A167359-50MG A167359-5G A167359-1G A167359-100MG
76973-836EA 51 USD
76973-836 76973-838 76973-840 77567-432 77881-292
2-Bromo-6-fluoropyrazine 97%
2-Bromo-6-fluoropyrazine

Precaución: For research use only.

Formula: C₄H₂BrFN₂
MDL Number: MFCD14702734
CAS Number: 1209458-12-7
UN: 2924
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR