(3-(4-Methylpiperazin-1-yl)phenyl)methanol 95%

Supplier: Ambeed
Warning

A129074-10G A129074-1G A129074-250MG
76971-966EA 545 USD
76971-966 76971-968 76971-970
(3-(4-Methylpiperazin-1-yl)phenyl)methanol 95%
(3-(4-Methylpiperazin-1-yl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD09757582
CAS Number: 123987-13-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR