Methyl-2-aminobenzo[d]thiazole-7-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-aminobenzo[d]thiazole-7-carboxylate

A260266-100MG A260266-1G A260266-250MG
76976-922EA 14.86 USD
76976-922 76976-924 76976-926
Methyl-2-aminobenzo[d]thiazole-7-carboxylate 98%
Methyl-2-aminobenzo[d]thiazole-7-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂S
MW: 208.2370 g/mol
MDL Number: MFCD09263762
CAS Number: 209459-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR