DL-Phenylalanine amide hydrochloride 99%

Supplier: Ambeed
Warning

A157079-10G A157079-1G A157079-25G A157079-5G
76973-354EA 43 USD
76973-354 76973-356 76973-358 76973-360
DL-Phenylalanine amide hydrochloride 99%
DL-Phenylalanine amide hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClN₂O
MDL Number: MFCD00070243
CAS Number: 108321-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR