2-Amino-N-methylpropanamide hydrochloride 96%

Supplier: Ambeed
Warning

A735282-1G A735282-5G
76981-312EA 47 USD
76981-312 76981-314
2-Amino-N-methylpropanamide hydrochloride 96%
2-Amino-N-methylpropanamide hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁ClN₂O
MDL Number: MFCD13562337
CAS Number: 38215-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR