3-Fluoro-2-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A204216-10G A204216-1G A204216-250MG A204216-25G
76975-312EA 132 USD
76975-312 76975-314 76975-316 76975-318
3-Fluoro-2-iodobenzoic acid 98%
3-Fluoro-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄FIO₂
MW: 266.0100 g/mol
MDL Number: MFCD07774249
CAS Number: 387-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR