(4-Methyl-1H-imidazol-5-yl)methanol hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxymethyl-5-methylimidazole hydrochloride

A124098-100G A124098-10G A124098-5G
76971-734EA 66.84 USD
76971-734 76971-736 76971-738
(4-Methyl-1H-imidazol-5-yl)methanol hydrochloride 95%
(4-Methyl-1H-imidazol-5-yl)methanol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₂O
MW: 148.5908 g/mol
MDL Number: MFCD00040986
CAS Number: 38585-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR