2-(2H-Benzo[d][1,2,3]triazol-2-yl)-4,6-di-tert-butylphenol 98%

Supplier: Ambeed
Warning

A165493-10G A165493-1G A165493-25G
76973-766EA 11.67 USD
76973-766 76973-768 76973-770
2-(2H-Benzo[d][1,2,3]triazol-2-yl)-4,6-di-tert-butylphenol 98%
2-(2H-Benzo[d][1,2,3]triazol-2-yl)-4,6-di-tert-butylphenol

Precaución: for Research Use Only

Formula: C₂₀H₂₅N₃O
MW: 323.43 g/mol
MDL Number: MFCD01075688
CAS Number: 3846-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR