L-Lysine ethyl ester dihydrochloride 98%

Supplier: Ambeed
Warning

A164403-100G A164403-10G A164403-25G A164403-500G
76973-718EA 66 USD
76973-718 76973-720 76973-722 76973-724 76973-726
L-Lysine ethyl ester dihydrochloride 98%
L-Lysine ethyl ester dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₂₀Cl₂N₂O₂
MDL Number: MFCD00039068
CAS Number: 3844-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR