Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A171932-100G A171932-25G
76974-036EA 27 USD
76974-036 77881-324
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid 98%
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MDL Number: MFCD00152680
CAS Number: 3813-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR