2-Amino-5-bromo-3-iodopyridine 96%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-iodo-2-pyridylamine, 5-Bromo-3-iodopyridin-2-amine

A279056-100G A279056-10G A279056-1G A279056-25G A279056-5G
76977-358EA 93 USD
76977-358 76977-360 76977-362 76977-364 77601-232
2-Amino-5-bromo-3-iodopyridine 96%
2-Amino-5-bromo-3-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₄BrIN₂
MDL Number: MFCD06659000
CAS Number: 381233-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR