5-Bromo-N,N-diethyl-2-pyridinamine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-N,N-diethylpyridin-2-amine

A586494-250MG A586494-1G
76980-424EA 130 USD
76980-424 77572-866
5-Bromo-N,N-diethyl-2-pyridinamine 95%
5-Bromo-N,N-diethyl-2-pyridinamine

Precaución: for Research Use Only

Formula: C₉H₁₃BrN₂
MDL Number: MFCD11938559
CAS Number: 1176424-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR