4-(Methylsulfonylamino)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Methylsulphonylamino)phenylboronic acid, 4-Methylsulphonylaminophenylboronic acid, 4-(Methanesulphonamido)phenylboronic acid

A596992-100MG A596992-1G A596992-250MG A596992-5G
76980-468EA 17 USD
76980-468 76980-470 76980-472 76980-474
4-(Methylsulfonylamino)benzeneboronic acid 98%
4-(Methylsulfonylamino)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄S
MDL Number: MFCD02179473
CAS Number: 380430-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR