4-Nitro-2,3-lutidine-N-oxide 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dimethyl-4-nitropyridine 1-oxide, 2,3-Dimethyl-4-nitropyridine N-oxide

A734212-100G A734212-25G A734212-500G A734212-5G
76981-296EA 51.99 USD
76981-296 76981-298 76981-300 76981-302
4-Nitro-2,3-lutidine-N-oxide 97%
4-Nitro-2,3-lutidine N-oxide

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 168.15 g/mol
MDL Number: MFCD00065172
CAS Number: 37699-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR