2-[4-(5-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(5-Aminopyridin-2-yl)piperazin-1-yl)ethanol

A369333-1G A369333-250MG
76978-688EA 159.14 USD
76978-688 76978-690
2-[4-(5-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol 95%
2-[4-(5-Amino-2-pyridinyl)-1-piperazinyl]-1-ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₈N₄O
MW: 222.29 g/mol
MDL Number: MFCD09882493
CAS Number: 1017221-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR