5-Cyano-2-fluoro-6-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-2-methylnicotinonitrile

A219280-10G A219280-1G A219280-250MG A219280-25G A219280-5G
76975-782EA 533.64 USD
76975-782 76975-784 76975-786 76975-788 76975-790
5-Cyano-2-fluoro-6-picoline 98%
5-Cyano-2-fluoro-6-picoline

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.1264 g/mol
MDL Number: MFCD03095096
CAS Number: 375368-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR