2,6-Pyridinedicarbonyl dichloride 95%

Supplier: Ambeed
Danger

Synonyms: 2,6-Pyridinedicarboxylicacidchloride, 2,6-Pyridinedicarboxylic acid chloride

A270699-1G A270699-25G A270699-5G
76977-158EA 11 USD
76977-158 76977-160 76977-162
2,6-Pyridinedicarbonyl dichloride 95%
2,6-Pyridinedicarbonyl dichloride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO₂
MW: 204.006 g/mol
MDL Number: MFCD00006289
CAS Number: 3739-94-4
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR