4,5-Diiodo-1-methylimidazole 98%

Supplier: Ambeed
Warning

A640366-250MG A640366-5G A640366-1G
76980-740EA 19 USD
76980-740 76980-742 77673-736
4,5-Diiodo-1-methylimidazole 98%
4,5-Diiodo-1-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₄I₂N₂
MDL Number: MFCD03001336
CAS Number: 37067-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR