4-Bromopyridin-2(1H)-one 99%
Supplier: Ambeed
|
Warning
|
A116321-100G
A116321-10G
A116321-1G
A116321-250MG
A116321-25G
A116321-500G
A116321-5G
76971-202EA
227
USD
76971-202
76971-204
76971-206
76971-208
76971-210
76971-212
76971-214
4-Bromopyridin-2(1H)-one 99%
4-Bromopyridin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₅H₄BrNO |
MDL Number:
MFCD11226860 CAS Number: 36953-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...