1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 95%

Supplier: Ambeed
Warning

A129247-10G A129247-25G A129247-5G
76971-978EA 100 USD
76971-978 76971-980 76971-982
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol 95%
1-[3,5-Bis(trifluoromethyl)phenyl]ethanol

Precaución: for Research Use Only

Formula: C₁₀H₈F₆O
MDL Number: MFCD01632521
CAS Number: 368-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR