5-Norbornene-2,3-dicarboximide 97%

Supplier: Ambeed
Warning

Synonyms: 3a,4,7,7a-Tetrahydro-1H-4,7-methanoisoindole-1,3(2H)-dione, Bicyclo[2.2.1]hept-5-ene-2,3-dicarboximide

A281757-100G A281757-10G A281757-25G A281757-5G
76977-426EA 127.31 USD
76977-426 76977-428 76977-430 76977-432
5-Norbornene-2,3-dicarboximide 97%
5-Norbornene-2,3-dicarboximide

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD08704199
CAS Number: 3647-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR