Methyl-5-amino-1H-1,2,4-triazole-3-carboxylate 95%

Supplier: Ambeed
Warning

A210903-100G A210903-10G A210903-1G A210903-25G A210903-500G A210903-5G
76975-536EA 107 USD
76975-536 76975-538 76975-540 76975-542 76975-544 76975-546
Methyl-5-amino-1H-1,2,4-triazole-3-carboxylate 95%
Methyl-5-amino-1H-1,2,4-triazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₄H₆N₄O₂
MDL Number: MFCD00297193
CAS Number: 3641-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR