2-Chloro-6-fluoroaniline 98%
Supplier: Ambeed
|
Warning
|
A233404-100G
A233404-10G
A233404-1G
A233404-25G
A233404-500G
A233404-5G
76976-220EA
149.59
USD
76976-220
76976-222
76976-224
76976-226
76976-228
76976-230
2-Chloro-6-fluoroaniline 98%
2-Chloro-6-fluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₅ClFN MW: 145.56 g/mol |
MDL Number:
MFCD00040309 CAS Number: 363-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...