1-tert-Butyl 2-ethyl piperidine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

A241070-100G A241070-10G A241070-25G A241070-500G A241070-5G
76976-456EA 76.39 USD
76976-456 76976-458 76976-460 76976-462 76976-464
1-tert-Butyl 2-ethyl piperidine-1,2-dicarboxylate 97%
1-tert-Butyl 2-ethyl piperidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₃NO₄
MW: 257.32 g/mol
MDL Number: MFCD03788452
CAS Number: 362703-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR