2-Chloro-4-methylbenzothiazole 98%

Supplier: Ambeed
Warning

A656297-1G A656297-5G
76980-840EA 25.47 USD
76980-840 76980-842
2-Chloro-4-methylbenzothiazole 98%
2-Chloro-4-methylbenzothiazole

Precaución: for Research Use Only

Formula: C₈H₆ClNS
MW: 183.658 g/mol
MDL Number: MFCD03840701
CAS Number: 3622-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR