2-Chloro-5-fluoro-4-methoxypyrimidine 98%

Supplier: Ambeed
Warning

A801906-10G A801906-1G A801906-250MG A801906-5G
76981-672EA 75 USD
76981-672 76981-674 76981-676 76981-678
2-Chloro-5-fluoro-4-methoxypyrimidine 98%
2-Chloro-5-fluoro-4-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₄ClFN₂O
MDL Number: MFCD07440062
CAS Number: 37554-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR