N-(Cyclobutylcarbonyl)-2-methylalanine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Cyclobutanecarboxamido)-2-methylpropanoic acid

A194030-250MG A194030-100MG
76974-964EA 53.05 USD
76974-964 77671-722
N-(Cyclobutylcarbonyl)-2-methylalanine 97%
N-(Cyclobutylcarbonyl)-2-methylalanine

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
MDL Number: MFCD13559827
CAS Number: 1219960-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR