2,2'-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol 97%

Supplier: Ambeed
Warning

Synonyms: N1,N4,N4-Tris(2-hydroxyethyl)-2-nitro-1,4-phenylenediamine

A186074-1G A186074-250MG
76974-624EA 9.55 USD
76974-624 76974-626
2,2'-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol 97%
2,2'-[4-(2-Hydroxyethylamino)-3-nitrophenylimino]diethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₉N₃O₅
MW: 285.30 g/mol
MDL Number: MFCD00071762
CAS Number: 33229-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR