5,5-Dimethyl-1-pyrroline-N-oxide 97%
Supplier: Ambeed
|
Warning
|
A189217-10G
A189217-1G
A189217-250MG
A189217-5G
A189217-25G
A189217-100G
76974-796EA
224
USD
76974-796
76974-798
76974-800
76974-802
77671-678
77881-409
5,5-Dimethyl-1-pyrroline-N-oxide 97%
5,5-Dimethyl-1-pyrroline-N-oxide
Precaución: for Research Use Only
Formula:
C₆H₁₁NO |
MDL Number:
MFCD00005279 CAS Number: 3317-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...