N-(tert-Butoxycarbonyl)-D-2-phenylglycine 98%
Supplier: Ambeed
|
Warning
|
A147204-100G
A147204-10G
A147204-25G
A147204-500G
A147204-5G
76972-884EA
45
USD
76972-884
76972-886
76972-888
76972-890
76972-892
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 98%
N-(tert-Butoxycarbonyl)-D-2-phenylglycine
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO₄ |
MDL Number:
MFCD00062043 CAS Number: 33125-05-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...