Chart Recorder Options for ULT Freezers
Supplier: Thermo Fisher Scientific
332166G01 332166G02 332791G01 332791G02
76779-124EA 1744.4 USD
76779-124 76779-126 76779-138 76779-140
Chart Recorder Options for ULT Freezers
Freezers Ultra Low Temperature Freezers
Chart recorder options for ultra-low temperature chest freezers.
  • Ink or inkless, physical record keeping for temperature tracking
Chart recorders provide an uneditable, 21 CFR Part 11 compliant, historical temperature recording for documentation and tracking.
Información para pedidos: Add to ordering configuration with ULT chest freezer.