N-Methoxy-N,2,3-trimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

90756-1G 90756-500MG 90756-5G
76944-130EA 548.49 USD
76944-130 76944-132 76944-134
N-Methoxy-N,2,3-trimethylbenzamide ≥97%
N-Methoxy-N,2,3-trimethylbenzamide
Formula: C₁₁H₁₅NO₂
Storage Temperature: Ambient
CAS Number: 229970-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR