5-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

89897-25G 89897-5G
76944-122EA 488.02 USD
76944-122 76944-124
5-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole ≥97%
5-Iodo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₈H₁₁IN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD19237232
CAS Number: 1311197-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR