Methyl 6-bromobenzo[d][1,3]dioxole-5-carboxylate ≥97%
Supplier: AOBChem USA
|
Warning
|
87624-1G
87624-25G
87624-500MG
87624-5G
76944-108EA
331.01
USD
76944-108
76944-110
76944-112
76944-114
Methyl 6-bromobenzo[d][1,3]dioxole-5-carboxylate ≥97%
Methyl 6-bromobenzo[d][1,3]dioxole-5-carboxylate
Formula:
C₉H₇BrO₄ MW: 259.053 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06203686 CAS Number: 61441-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...